missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Slotted Gaskets for SE 400 and SE 600 Series Units
Slotted Gaskets for SE 400 and SE 600 Series Units
POA
Specifications
Color | Gray |
---|---|
For Use With (Equipment) | SE 400, SE 600 Series Vertical Electrophoresis Unit |
Type | Slotted Gasket |
Material | Silicone Rubber |
Description
- For use with SE 400 and SE 600 Series Units
Specifications
Gray | |
SE 400, SE 600 Series Vertical Electrophoresis Unit | |
Slotted Gasket | |
Silicone Rubber |