Learn More
Hoefer™ SG Series Gradient Makers
Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride
Brand:  Hoefer™ SG500
Code : 04
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 11574424
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Models SG 15, SG 30, SG 50, and SG 100
  • Milled of heavy acrylic plastic with high-quality, leak-free PTFE-resin valves
  • Offer a flat base for stability on magnetic stirrers; support rod can be attached to ring stand for additional stability
  • Accept Luer or 4mm ID tubing; with included adapters can also accept 2mm ID or 22-gauge tubing
  • SG 100 is ideal for the Hoefer Mighty Small™ Gel Casters SE 275 (for four gels) or SE 215 (for 10 gels)
Model SG 500
  • Made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base
  • Uses a simple push-pull valve to open and close the passage between the reservoir and mixing chambers
  • Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns
  • Ideal for use with the Hoefer SE 615 or SE 915 Multiple Gel Casters
Specifications
Specifications
SE 615, SE 915 Multiple gel casters
Gradient Maker
Gradient maker w/Push-Pull Valve, four wrap-and-seal hose clamps
500mL
Videos
Documentation
Technical Documents
View accessory products