missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SE 915 Multicaster

Suitable for use with the SE 900 Large Format Vertical Electrophoresis System as part of the 2-Dimensional Electrophoresis System

Brand:  Hoefer™ SE915

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11711879

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Requires : Cassettes sold separately.

Specifications

Specifications

0.1cm
28 x 21cm Cassette
Multicaster
One Acrylic space saver, one filler plug and ten divider sheets
Videos
Documentation View accessory products