Learn More
Cytiva SE 400 Vertical Unit
The sturdier vertical slab gel unit was designed with durability and cost in mind. Injection-moulded of tough, impact-resistant plastics, it will withstand the rigours of daily lab use and teaching labs for years
Brand: Cytiva SE400-15-1.5
Description
- SE 400 Vertical Units run one 16-cm gel or two gels with an accessory divider plate.
- Its low cost makes the SE 400 appropriate for labs requiring multiple units when temperature control is not crucial.
- The unit base is also a convenient casting stand for single gels. To cast multiple gels for one or more units, use the multiple gel casters.
- Buffer volume: 350 ml (Minimum upper buffer volume) 150 ml (Minimum lower buffer volume) 350 ml (Maximum lower buffer volume)
- Total Separation Time: 3-5 h
Specifications
One set of glass plates 18 x 16 cm, 2 clamp assemblies and 2 cams, 15-well comb and 2 spacers 1.5 mm thick | |
3.2 kg | |
<20 w |
0°C to 45°C | |
500 VDC |