missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, T-Spacers

For use with SE 250 vertical electrophoresis unit

Brand:  Hoefer™ SE2119T-2-1.0

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12054847

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

8cm
0.1cm
SE 250 vertical electrophoresis unit
PVC
0.1cm
White
T-shape spacer
Videos
Documentation

Technical Documents