Learn More
Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates
Use with the SE 250 vertical electrophoresis unit. Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
Brand:  Hoefer™ SE202P-10
Code : 04
Additional Details : Weight : 0.28200kg
 View more versions of this product
Product Code. 11519653
  • £71.55 / Pack of 10
In Stock 4
Add to basket
You may only order up to the available stock. For more details call customer services on 01509 555 500
Description
Description
  • 10 x 8cm
Specifications
Specifications
SE 250 vertical electrophoresis unit
Glass
10/Pk.
Low Fluorescence Glass Plate
Rectangular
Videos
Documentation
Technical Documents