Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates
Use with the SE 250 vertical electrophoresis unit. Hoefer™ SE 250 Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
Overview
Brand: Hoefer™
Manufacturer Part Number: 80613624
UNSPSC: 41105305
Code: GH
Additional Details:
Additional Details: Weight: 0.03200kg
Product Code. 12981570
Quantity Price
1 £ 38.24 / Each
Estimated Shipment
18-04-2017
Add to basket
If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.
Description and Specification
Specification
Product Type Notched Alumina Plate
Material Alumina
For Use With (Equipment) SE 250 vertical electrophoresis unit
  • 10 x 8cm