missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems

Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

Brand:  Hoefer™ SE260B

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11741789

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Includes: SE250 Basic System includes: upper buffer chamber, lower buffer chamber, interlocking safety lid with leads, ten 10 x 8cm glass plates, two 10 x 8cm notched alumina plates, four spring clamps, gel seal, well locating decal, and Spacer Mate. SE250 Complete System also includes: Dual Gel Caster, two 0.75mm thick 10-well combs, and two pairs 0.75mm thick spacers. SE260 Basic System includes same items as SE250 Basic Systems, except the glass and alumina plates.SE260 Complete Systems add Dual Gel Caster, ten 10.5 x 10cm glass plates, five 10.5 x 10cm notched alumina plates, and two 10-well combs and two pairs spacers (either 0.75mm or 1.5mm thick; see ordering table).

Description

Description

  • Run two gels at once
  • Produce high-resolution separations in as little as 45 minutes with no band “smiling”
  • Use alumina plates (included with specified models) for efficient cooling
  • Buffer volume: 300mL

Applications:

Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis

Certifications

CE Marked, CSA C22.2 1010.1, EN61010-1, UL3101-1.

Specifications

Specifications

500mA
Up to 80%
45°C
500V
300mL
10cm
16.5cm
16cm
Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, four spring clamps, gel seal, well locating decal and spacer mate
15° to 30°C
12w
8/10.5cm
18cm
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Videos
Documentation