missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ HE 33 Mini Submarine Electrophoresis Unit

Designed for very fast separations of DNA restriction fragments in agarose gels

Brand:  Hoefer™ HE33B

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12074657

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
  • Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
  • No taping required for casting
  • Electrical safety locks in lid

Specifications

Specifications

500mA
Up to 80%
Up to 32
500V
Mini horizontal Agarose electrophoresis unit
10cm
7cm
13cm
9.4 x 5.1 x 2.8 in. (24 x 13 x 7cm)
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level
50°C
15w
250mL
7cm
24cm
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Videos
Documentation

Technical Documents

View accessory products