missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Glass Plates for Vertical Electrophoresis Units

Glass plates for vertical electrophoresis units

Brand:  Hoefer™ SE6602

Code : GE

Additional Details : Weight : 0.79000kg

 View more versions of this product

Product Code. 10630796

  • £161.00 / Pack of 2


Estimated Shipment
19-07-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

  • For use with Hoefer SE600 and SE400 Series Vertical Electrophoresis Units
  • Storage temperature: +15°C to +30°C

Specifications

Specifications

SE 600, SE 400 series vertical electrophoresis system
Glass Plate
Glass
Videos