missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Gel Casting Accessories for HE 99X Max Submarine Units: Casting Kits

For use with Hoefer HE 99X Max Submarine Electrophoresis Units

Brand:  Hoefer™ HE97X-20

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11599563

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Casting kits include necessary components to cast agarose gels without tape—includes casting tray, running tray, and foam gaskets

Specifications

Specifications

20cm
Casting Kit
Running tray, casting tray, Foam gasket
15 x 20cm
15cm
HE99X max submarine electrophoresis unit
7.8 x 5.9 in. (20 x 15cm)
Videos
Documentation

Technical Documents