Learn More
Hoefer™ Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
Brand: Hoefer™ SE275
Code : 04
Additional Details : Weight : 0.80000kg
Description
SE245 Dual-Gel Caster
- For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
- Suitable for single concentration and gradient gels
- SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
- SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
Specifications
SE 250 Mini Vertical Gel Electrophoresis Unit | |
Gel Caster | |
Casting cradle with gasket, face, Glass, alumina, space saver plates, filler sheets, plugs | |
1 to 4 |