missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Brand:  Hoefer™ SE511-10-1.5

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13127814

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Lightweight
  • Virtually unbreakable

  • Come attached to a standard back which produces maximum well depths of 25mm, except for the 28-well preparative combs which produce well depths of 15mm
  • Adjustable comb back (available separately, below) converts 25mm deep combs to 10 or 15mm depth

Specifications

Specifications

0.15cm
12.4μL/mm
SE 600, SE 400 series vertical electrophoresis system
10
0.83cm
2.5cm
Comb
PTFE
Videos
Documentation

Technical Documents