missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Brand:  Hoefer™ SE511-BKA

Code : GE

Additional Details : Weight : 0.01400kg

 View more versions of this product

Product Code. 10783847

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications

Specifications

SE 600, SE 400 Series Vertical Electrophoresis System
PTFE
2.5 cm
Adjustable Comb Back
0.98 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers