missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand: Hoefer™ SE211A-18-1.0
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Lightweight
- Virtually unbreakable
Specifications
0.1cm | |
2.9μL/mm | |
SE 250, SE 260, MiniVE™ vertical electrophoresis system | |
18 |
0.29cm | |
1.3cm | |
Comb | |
PTFE |