missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Brand:  Hoefer™ SE211A-10-1.0

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12094797

  • £43.80 / Each


Estimated Shipment
22-03-2018
Add to basket

Specifications

Specifications

0.1cm
4.8μL/mm
SE 250, SE 260, MiniVE™ vertical electrophoresis system
10
0.48cm
1.3cm
Comb
PTFE
Videos
Documentation

Technical Documents