Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand:  Hoefer™ SE211A-15-1.5
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12004707
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Lightweight
  • Virtually unbreakable
Specifications
Specifications
0.15cm
4.4μL/mm
SE 250, SE 260, MiniVE™ vertical electrophoresis system
15
0.29cm
1.3cm
Comb
PTFE
Videos
Documentation
Technical Documents