Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand: Hoefer™ SE211A-10-.75
Code : 04
Additional Details : Weight : 1.00000kg
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System | |
PTFE | |
0.03 in. | |
9.8μL/ mm | |
0.51 in. |
Comb | |
10 | |
0.075 cm | |
0.48 cm | |
1.3 cm |