missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Brand:  Hoefer™ SE211A-R-.75

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12059807

  • £43.80 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Lightweight
  • Virtually unbreakable

Specifications

Specifications

0.07cm
1.3cm
Comb
PTFE
51μL/mm
SE 250, SE 260, MiniVE™ vertical electrophoresis system
1 Preparative, 1 Reference
6.8cm
Videos
Documentation

Technical Documents