missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit

Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit

Brand:  Hoefer™ HE91A-30-1.0

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12014737

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Requires : Comb back must be ordered separately (see ordering table)

Specifications

Specifications

0.1cm
Backless Comb
30
0.3cm
0.5cm
HE99X max submarine electrophoresis unit
15μL
Videos
Documentation

Technical Documents

View accessory products