missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

Brand:  Hoefer™ 80614859

Code : GH

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12961590

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance



SE 250
Buffer Chamber