missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

Brand:  Hoefer™ SE255D

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12054737

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

SE 260
Buffer Chamber
Videos
Documentation

Technical Documents