missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1 Monoclonal Antibody (6B8-2F2), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA545347
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45347 was sufficient for detection of HO-1 in 10 μg of rat kidney lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.∣Detects approximately 32kDa. Does not cross-react with HO-2.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.Specifications
HO-1 | |
Monoclonal | |
1 mg/mL | |
9.1mM sodium bicarbonate, 9.44mM PBS with 123mM ethanolamine, 581mM DMSO and no preservative | |
P06762, P09601, P14901 | |
HMOX1 | |
His-tagged Rat HO-1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Western Blot | |
6B8-2F2 | |
FITC | |
HMOX1 | |
bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein | |
Mouse | |
Protein G | |
RUO | |
15368, 24451, 3162 | |
4° C, store in dark | |
IgG1 κ |