missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPA0 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557722
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RGFGFVYFQN HDAADKAAVV KFHPIQGHRV EVKKAVPKED IYSGG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
HNRNPA0 Polyclonal antibody specifically detects HNRNPA0 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HNRNPA0 | |
Polyclonal | |
Unconjugated | |
Hnrnpa0 | |
1110055B05Rik; 3010025E17Rik; Heterogeneous nuclear ribonucleoprotein A0; hnRNA binding protein; hnRNP A0; HNRNPA0; Hnrpa0 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10949, 498696, 77134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13151, Q9CX86 | |
Hnrnpa0 | |
Recombinant protein corresponding to Human HNRNPA0 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only