missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP U Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559411
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TEQKGGDKKR GVKRPREDHG RGYFEYIEEN KYSRAKSPQP PVEEEDEHFD DTVVCLDT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.
hnRNP U Polyclonal antibody specifically detects hnRNP U in Human samples. It is validated for Immunohistochemistry.Specifications
hnRNP U | |
Polyclonal | |
Unconjugated | |
Hnrnpu | |
AA408410; AI256620; AL024194; AL024437; AW557595; C1orf199; C86794; FLJ30202; FLJ37978; GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; hnRNP U; HNRNPU; HNRNPU antisense RNA 1; HNRNPU-AS1; Hnrpu; NCRNA00201; nuclear matrix protein sp120; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; rCG_20317; SAFA; SAF-A; scaffold attachment factor A; Scaffold-attachment factor A; SN1; SP120; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U21.1; U211 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3192 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q00839 | |
Hnrnpu | |
Recombinant protein corresponding to Human hnRNP U | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only