missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP DL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564094
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HVTAQQPSRL AGGAAIKGGR RRRPDLFRRH FKSSSIQRSA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
hnRNP DL Polyclonal antibody specifically detects hnRNP DL in Human samples. It is validated for Immunohistochemistry.Specifications
hnRNP DL | |
Polyclonal | |
Unconjugated | |
HNRNPDL | |
A+U-rich element RNA binding factor; AA407431; AA959857; AU-rich element RNA-binding factor; D5Ertd650e; D5Wsu145e; heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa); heterogeneous nuclear ribonucleoprotein D like; heterogeneous nuclear ribonucleoprotein D-like; heterogenous ribonucleoprotein D-like; HNRNP; hnRNP DL; hnRNP D-like; HNRNPD; Hnrnpdl; hnRNP-DL; Hnrnpdl1; HNRPDL; hnRPD-like protein; JKT41-binding protein; JKTBP; JKTBP2; laAUF1; LGMD1G; LOC534770 protein; protein laAUF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14979 | |
HNRNPDL | |
Recombinant protein corresponding to Human hnRNP DL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only