Learn More
HNMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557288
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TSDDLTQMLD NLGLKYECYD LLSTMDISDC FIDGNENGDL LWDFLTETCN FNATAPPDLR AELGKDLQEP EFSAKKEGKV LFNNTLSFIV IEA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
HNMT Polyclonal antibody specifically detects HNMT in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
HNMT | |
Polyclonal | |
Unconjugated | |
Hnmt | |
1500031F01Rik; AI788969; histamine N-methyltransferase; Hmt; Hnmt; HNMT-S1; HNMT-S2; MRT51 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P50135 | |
Hnmt | |
Recombinant protein corresponding to Human HNMT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only