missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555620
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ILQLTRAVQE TSLTPARLLP VAHQRFSTAS AVPLAKTDTW PKDVGILALE VYFPAQYVDQ TDLEKYNNVE Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
HMGCS2 Polyclonal antibody specifically detects HMGCS2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HMGCS2 | |
Polyclonal | |
Unconjugated | |
HMGCS2 | |
1300002P16; 3-hydroxy-3-methylglutary-Coenzyme A synthase 2; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 2; 3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial); 3-hydroxy-3-methylglutaryl-coenzyme A synthase; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 2 (mitochondrial); hmgCoA; HMG-CoA synthase; Hmgcs1; Hmgcs2; hydroxymethylglutaryl-CoA synthase 2; hydroxymethylglutaryl-CoA synthase, mitochondrial; mHS; Mt3h3mg; testicular tissue protein Li 88 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P54868 | |
HMGCS2 | |
Recombinant protein corresponding to Human HMGCS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only