Learn More
HMGCR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552547
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MAGSIGGYNA HAANIVTAIY IACGQDAAQN VGSSNCITLM EASGPTNEDL YISCTMPSIE IGTVGGGTNL LPQQACLQML GVQGACKDNP GENARQLARI VCGTVMAGEL SLMAALAAGH LVKSHMIHNR SKINLQDLQG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
HMGCR Polyclonal antibody specifically detects HMGCR in Human samples. It is validated for Immunohistochemistry.Specifications
HMGCR | |
Polyclonal | |
Unconjugated | |
HMGCR | |
3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3156 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P04035 | |
HMGCR | |
Recombinant protein corresponding to Human HMGCR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only