Learn More
HMBOX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565983
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: IKRRANIEAAI LESHGIDVQS PGGHSNSDDV DGNDYSEQDD STSHSDHQDP ISLAVEMAAV NHTILALARQ GANEIKTEAL DD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 47%.
HMBOX1 is located at the boundary of 8p12.3 and 8p21.1. HMBOX1 proteins are highly conserved in human, mouse, rat, chicken and Xenopus laevis. Functional HMBOX1::EGFP (enhanced green fluorescent protein) fusion protein revealed that HMBOX1 accumulated more in cytoplasm than in nucleus. HMBOX1 is a transcription repressor. Human HMBOX1 is widely expressed in 18 tissues, and it is highly expressed in pancreas. Hmbox1 is widely expressed in mouse pancreas and the expression of this gene can also be detected in pallium, hippocampus and hypothalamus.Specifications
HMBOX1 | |
Polyclonal | |
Unconjugated | |
HMBOX1 | |
AI451877; AI604847; F830020C16Rik; HMBOX1; HNF1LA; homeobox containing 1; homeobox telomere-binding protein 1; Homeobox-containing protein 1; homeobox-containing protein PBHNF; HOT1; PBHNF; RGD1306787; TAH1; Telomere-associated homeobox-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79618 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NT76 | |
HMBOX1 | |
Recombinant Human HMBOX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only