missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HM13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565995
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: KGEVTEMFSYE ESNPKDPAAV TESKEGTEAS ASKGLEKKEK Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
HM13 Polyclonal antibody specifically detects HM13 in Human samples. It is validated for Immunocytochemistry.Specifications
HM13 | |
Polyclonal | |
Unconjugated | |
HM13 | |
1200006O09Rik; 4930443L17Rik; 5031424B04Rik; AV020344; cb228; H13; H-13; hIMP1; histocompatibility (minor) 13; histocompatibility 13; histocompatibility minor 13; HM13; hm13 protein; IMP1; IMP-1; IMPAS; IMPAS-1; intramembrane aspartyl protease; intramembrane protease 1; minor histocompatibility antigen 13; minor histocompatibility antigen H13; MSTP086; presenilin-like aspartyl protease; presenilin-like protein 3; PSENL3; PSL3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1; SPP; SPPL1; Unknown (protein for MGC:134054); wu:fc11a06; zgc:56660; zgc:86862 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81502 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TCT9 | |
HM13 | |
Recombinant Human HM13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only