missing translation for 'onlineSavingsMsg'
Learn More

HLA-E, clone: 3D12HLA-E, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9953-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288267

  • £68.39 / 25µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody 3D12 (HLA-E) recognizes human HLA-E, a member of the Human Leukocyte Antigen family. HLA-E is a MHC Class IB member, which has a specialized role in NK cell recognition. Binding of a restricted set of peptides derived from other Class I leader sequences to the HLA-E and B2M (beta2 microglobulin) complex leads to interaction with NKG2A on NK cells. This binding protects the HLA-G-expressing cell from NK cell killing. HLA-E expression is ubiquitous.Applications Tested: This 3D12 (HLA-E) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-E
3D12HLA-E
Unconjugated
Purified
IgG1
RUO
Primary
Human
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C.
Monoclonal
Videos