missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR, FITC, clone: LN3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9956-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15516656

  • £135.24 / 100 tests


EU Stock 3
Add to basket

Description

Description

Description: The LN3 mAb reacts with the human major histocompatibility complex (MHC) class II, HLA-DR. HLA-DR is expressed on the surface of human antigen presenting cells (APC) including B cells, monocytes, macrophages, DCs, and activated T cells. HLA-DR is a heterodimeric transmembrane protein composed of alpha and beta subunits and plays an important role in the presentation of peptides to CD4+ T lymphocytes.Applications Reported: This LN3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This LN3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-DR
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01903
XXbac-BPG161M6.1, DRB1, DRw10, FLJ75017, FLJ76359, HLA-DR1B, HLA-DRB, HLA-DRB1*, SS1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
LN3
FITC
HLA-DRA
Conjugated
HLA-DRA
IgG2b, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3122
Videos

For Research Use Only.