missing translation for 'onlineSavingsMsg'
Learn More

HLA-ABC, FITC, clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9983-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506666

  • £111.88 / 100 tests


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, C. MHC class I antigens associated with beta 2-microglobulin are expressed by all human nucleated cells and are central in cell-mediated immune response and tumor surveillance. W6/32 mAb recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both 43kDa and 11-12kDa chains. Crossreactivity is also seen in baboon, rhesus and cynomolgus monkey.Applications Reported: The W6/32 antibody has been reported for use in flow cytometric analysis.Applications Tested: This W6/32 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5μL (1μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-ABC
W6/32
FITC
Conjugated
IgG2a
RUO
Primary
Human
Flow Cytometry
5μL (1.0μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos