missing translation for 'onlineSavingsMsg'
Learn More

HLA-ABC, Biotin, clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9983-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15390140

  • £49.65 / 25µg


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, C. MHC class I antigens associated with beta 2-microglobulin are expressed by all human nucleated cells and are central in cell-mediated immune response and tumor surveillance. W6/32 mAb recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both 43kDa and 11-12kDa chains. Crossreactivity is also seen in baboon, rhesus and cynomolgus monkey.Applications Reported: This W6/32 antibody has been reported for use in flow cytometric analysis.Applications Tested: This W6/32 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-ABC
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P05534
H-2K, H-2K(d), H2-K, MGC184092, MGC7052
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
W6/32
Biotin
HLA-A
Conjugated
HLA-A
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
3105
Videos

For Research Use Only.