missing translation for 'onlineSavingsMsg'
Learn More

HLA-ABC, Biotin, clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9983-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300150

  • £111.93 / 100µg


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, C. MHC class I antigens associated with beta 2-microglobulin are expressed by all human nucleated cells and are central in cell-mediated immune response and tumor surveillance. W6/32 mAb recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both 43kDa and 11-12kDa chains. Crossreactivity is also seen in baboon, rhesus and cynomolgus monkey.Applications Reported: This W6/32 antibody has been reported for use in flow cytometric analysis.Applications Tested: This W6/32 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-ABC
W6/32
Biotin
Conjugated
IgG2a
RUO
Primary
Human
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos