missing translation for 'onlineSavingsMsg'
Learn More

HLA-A2, PerCP-eFluor™ 710, clone: BB7.2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9876-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15569456

  • £88.75 / 25 tests

Estimated Shipment
01-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells.The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28.Applications Reported: This BB7.2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This BB7.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-A2
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P05534
DAQB-90C11.16, FLJ26655, HLAA
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BB7.2
PerCP-eFluor 710
HLA-A
Liquid
HLA-A
IgG2b, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3105
Videos
Documents

For Research Use Only.