missing translation for 'onlineSavingsMsg'
Learn More

HLA-A2, PE-Cyanine7, clone: BB7.2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-9876-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15528686

  • £74.73 / 25 tests


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells.The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28.Applications Reported: This BB7.2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This BB7.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HLA-A2
BB7.2
PE-Cyanine7
Conjugated
IgG2b
RUO
Primary
Human
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Videos