Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
Brand:  Simport™ Scientific M492-12
Code : 04
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10131011
  • £192.00 / Pack of 1500
Estimated Shipment: 22-11-2021
Description
Description
  • Made of acetal
  • Keeps specimens safely submerged in liquid
  • Resistant to the chemical action of most histological solvents
  • 45° angle anterior writing area
  • One-piece integral lid eliminates need for separate steel lids
  • Back mounted tab allows for an unobstructed anterior marking area
  • Open and close as often as necessary — cassettes will always relock securely
  • Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes
Certifications
CE Marked
Specifications
Specifications
Yes
Tissue Cassette
Aqua
Acetal
Videos
SDS
Documents
Certificates
For in vitro diagnostic (IVD) use only.