Learn More
Simport™ Histosette™ I Biopsy Processing and Embedding Cassettes
Specifically designed to hold biopsy specimens during the embedding process, as well as in a storage file.
Brand:  Simport™ M491-3
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10180671
Quantity Price
1 £63.54 / Pack of 1500
10 £57.07 / Pack of 1500
Estimated Shipment: 07-12-2021
Description
Description
  • Made of acetal
  • Disposable plastic cassettes
  • Anterior writing area at a 30° angle
  • Not suitable for automated printers
  • Each case contains 3 dispenser boxes of 500 cassettes
Specifications
Specifications
1500/Cs
Pink
Biopsy/embedding cassette
30°
Acetal