missing translation for 'onlineSavingsMsg'
Learn More

Histone H1 Rabbit, Clone: HH1/1784R, enQuireBio™

Rabbit Monoclonal Antibody

Brand:  EnquireBio 3005-RBM4-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15970158

  • £431.40 / 100µg
Estimated Shipment: 21-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Histone H1
Nuclear
200μg/mL
10mM PBS with or without 0.05% BSA and 0.05% azide.
H1
Recombinant full-length human Histone H1 protein
∽30kDa
Protein A/G Purified
RUO
Primary
3005
Immunohistochemistry
HH1/1784R
Unconjugated
NP_005309
Rabbit
IgG kappa
HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma.
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates
Special Offers

Special Offers