missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2BA Antibody (OTI1C9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72479AF405
This item is not returnable.
View return policy
Description
HIST1H2BA Monoclonal specifically detects HIST1H2BA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HIST1H2BA | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HIST1H2BA | |
| Recombinant protein fragment corresponding to amino acids 2-127 of human HIST1H2BA (NP_733759) produced in E.coli. | |
| 0.1 mL | |
| 255626.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1C9 | |
| Western Blot, Immunohistochemistry | |
| histone H2B type 1-A, bA317E16.3, H2BFU, histone cluster 1, H2ba, STBP, TSH2B | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction