missing translation for 'onlineSavingsMsg'
Learn More

6x-His Tag, clone: HIS.H8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6657-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537466

  • £69.10 / 25µg


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This HIS.H8 monoclonal antibody reacts with the 6X polyhistidine (6X His) tag. The H-H-H-H-H-H motif is frequently fused to the N- or C-terminus of proteins to facilitate purification (e.g., nickel affinity chromatography and immunoprecipitation) and/or detection. This antibody recognizes the 6X His tag anywhere within the protein of interest (i.e., N-terminus, C-terminus, or internal).Applications Reported: This HIS.H8 antibody has been reported for use in immunoprecipitation and western blotting.Applications Tested: This HIS.H8 antibody has been tested by immunoblotting of reduced cell lysate expressing a His-tagged protein at less than or equal to 1 ug/ mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

His Tag
HIS.H8
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
6X His tag
IgG2b
RUO
Primary
Murine
Videos