missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK4 Recombinant Human Protein
HIPK4 Recombinant Human Protein
Brand: Invitrogen PV3852
Code : 50
Additional Details : Weight : 0.05250kg
Description
HIPK4 is a homeodomain-interacting protein kinase and has been demonstrated to phosphorylate p53.Specifications
NP_653286 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
96.9kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
HIPK4 | |
HIPK4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
HIPK4 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.42mg/mL | |
1387, 147746 | |
HIPK4 Recombinant Human Protein | |
–80°C | |
1 Each | |
HIPK4 Recombinant Human Protein | |
DYRK | |
81nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.