missing translation for 'onlineSavingsMsg'
Learn More

High temperature urethane, U-835, Thermo Scientific™

Catalog Number 15492757
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Recommended for use as a base resin or additive for adhesives designed for metal substrates
TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Yellow liquid
Density 0.99 g/mL
Quantity 25 g
Flash Point >200°C (392°F)
Chemical Name or Material High temperature urethane, U-835
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'