missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ High-Impact Polystyrene Trays

Suitable for food stuff and general laboratory use.

Brand:  Kartell™ 5709

Code : 02

Additional Details : Weight : 0.52500kg

 View more versions of this product

Product Code. 11341205

  • £18.70 / Each


In Stock 18
Add to basket

Description

Description

  • With ridged feet for stability
  • Not autoclavable
Specifications

Specifications

1.6L
Ext.: 202 x 151 x 81mm; Int.: 140 x 90 x 70mm
Polystyrene
No
White
20/Pack
Tray
Foodstuffs and general laboratory use
Videos
Documentation