missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ High-Impact Polystyrene Trays

Suitable for food stuff and general laboratory use.

Brand:  Kartell™ 5702

Code : 02

Additional Details : Weight : 0.10340kg

 View more versions of this product

Product Code. 11361245

  • £18.54 / Each


In Stock 21
Add to basket

Description

Description

  • With ridged feet for stability
  • Not autoclavable
Specifications

Specifications

0.7L
Ext.: 303 x 151 x 21mm; Int.: 270 x 120 x 10mm
Polystyrene
No
White
1/Pack
Tray
Foodstuffs and general laboratory use
Videos
Documentation