missing translation for 'onlineSavingsMsg'
Learn More

HIF-1 alpha, clone: ESEE122, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9100-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258147

  • £69.66 / 25µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This ESEE122 monoclonal antibody reacts with hypoxia inducible factor-1 alpha. The hypoxia inducible factor (HIF-1) is a transcription factor composed of an inducible alpha form and the constitutively expressed beta form, also called the aryl hydrocarbon receptor nuclear translocator (ARNT). Both alpha and beta subunits of HIF-1 contain basic helix-loop-helix motifs and function to activate transcription of genes in response to reduced oxygen levels. Under normoxic conditions, HIF-1 alpha is rapidly degraded, while HIF-1 alpha expression is induced and degradation inhibited under hypoxic conditions. HIF-1 alpha can be found in the cytoplasm and/or in the nucleus under normoxic conditions, but translocates to the nucleus under hypoxic conditions. HIF-1 is upregulated in several cancer types and functions to control genes involved in angiogenesis, cell survival and T cell development. Recent studies have shown the importance of HIF-1 alpha in balancing Th17 and Treg development.This ESEE122 antibody has also been reported to cross-react with bovine, rat, and mouse hypoxia inducible factor-1 alpha.Applications Reported: This ESEE122 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.Applications Tested: This ESEE122 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded (FFPE) human placenta at less than or equal to 10 ug/mL using low pH antigen retrieval buffer. This ESEE122 antibody has been tested by immunocytochemistry on fixed and permeabilized Hela cells at less than or equal to 10 ug/mL. It is recommended that the antibody be titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HIF-1
ESEE122
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry (Formalin/Paraffin)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ARNT interacting protein, hypoxia inducible factor-1
IgG1
RUO
Primary
Rat
Videos