Learn More
HF scientific Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Brand:  HF scientific 10504
UNSPSC : 41104200
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13171491
  • £30.56 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
Hach™ and Hanna photometers
100.0
Dispenser
10mL
For testing free chlorine
Documentation