HF scientific Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Overview
Brand: HF scientific
Manufacturer Part Number: 10500
UNSPSC: 41115311
Code: 03
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 13131491
Quantity Price
1 £ 22.92 / Each
Estimated Shipment
27-04-2017
Add to basket
Description and Specification
Specification
Product Type Dispenser
For Use With (Application) For testing free chlorine
Sample Size 5mL
No. of Tests 100.0
For Use With (Equipment) HF scientific and Hach™ photometers
Documentation